Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Synthesis of Aspirin
GREENER_CHOICESCI_ED

Catalog No. s90746 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S90746 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid and Toluene, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S90746 Supplier Innovating Science™ Supplier No. IS8021
Only null left
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid and Toluene, which are known to the State of California to cause cancer and reproductive harm.

Includes

Materials for 15 groups: 25mL Acetic Anhydride UN1715 (2); 15g Salicylic Acid; 25mL Sulfuric Acid concentrate UN1830 (2); 25mL Toluene UN1294; teacher's guide; and student copy masters

Performs the synthesis of acetylsalicylic acid (aspirin) purification by recrystallization and using melting point to confirm identity.

In a reaction vessel, salicylic acid, acetic anhydride and sulfuric acid are mixed. The exothermic reaction will cause the temperature to increase to 70°-80°C. Once the reaction is complete, the vessel is cooled in an ice bath and the acetylsalicylic crystallizes out. This material is then recrystallized in toluene to purify the product. The product identity is comfirmed by melting point. The mass of the recrystallized product is used to determine the yield of the synthesis.

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Performs the synthesis of acetylsalicylic acid purification by recrystallization and using melting point to confirm identity, the mass of the recrystallized product is used to determine the yield of the synthesis
Includes 2 x 25ml acetic anhydride, 1 x 15g salicylic acid, 2 x 25ml sulfuric acid concentrate, 2 x 25ml toluene, teacher's guide and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 6.75 x 5 x 6 in.
Class Size 15 Lab Groups
Product Type Synthesis of Aspirin Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.