Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: The Electrochemical Series
GREENER_CHOICESCI_ED

Catalog No. S96899 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S96899 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96899 Supplier Innovating Science™ Supplier No. IS8030
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Materials for 15 groups: EZ-Prep™ to make 1000 mL of 1.0M solution of Copper Sulfate, Zinc Sulfate, Magnesium Sulfate, Iron Sulfate and Aluminum Sulfate Copper (15 strips); Copper (15 strips); Zinc (15 strips); Magnesium (15 strips); Iron (15 strips); Aluminum (15 strips); filter paper strips (75); teacher's guide; and student copy masters

Discover electrode potential.

  • The electrochemical series is built up by arranging various redox equilibria in order of their standard electrode potentials When a strip of metal is placed in water, the metal has a tendency to go into solution as ions with a simultaneous buildup of electrons on the metal strip
  • The process produces an electrical potential difference between the metal and solution

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Students will study electrochemical series, electrochemical series is built up by arranging various redox equilibrium in order of their standard electrode potentials
Includes 15 Copper Metal strips, 15 Zinc Metal strips, 15 Magnesium Metal strips, 15 Iron Metal strips, 15 Aluminum Metal strips, 75 filter Paper strips, EZ-Prep to make 1000 mL of 0.1M solution of Copper Sulfate, Zinc Sulfate, Magnesium Sulfate, Iron Sulfate, Alum
Quantity 1
Dimensions (L x W x H) 9 x 6 x 4 in.
Class Size 15 Lab Groups
Product Type The Electrochemical Series
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.