Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: The Molar Volume of a Gas
SCI_ED

Catalog No. S97013 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S97013 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S97013 Supplier Innovating Science™ Supplier No. IS8026
Add to Cart
Add to Cart

Includes

Materials for 15 groups: 25g Potassium Chloride UN1485 (2); 25g Manganese Dioxide, 85% native powder UN1479 (2); teachers guide; and student copy masters

Determines the volume of one mole of gas.

  • Generates a known mass of oxygen gas
  • Measures the temperature, volume and pressure
  • Uses data to calculate molar volume at STP
  • Determines the percentage KClO3 in an unknown sample

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To determine the volume of one mole of gas
Includes 2 x 25g potassium chlorate, 2 x 25g manganese dioxide, 85% native powder, Teacher's guide and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 9.75 x 6.75 x 3 in.
Class Size 15 Lab Groups
Product Type Molar Volume of A Gas Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.