Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Thermochemistry
SCI_ED

Catalog No. S90726 Shop All Aldon Products
Change view
Click to view available options
AP Chemistry Kit: Thermochemistry
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S90726 AP Chemistry Kit: Thermochemistry
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S90726 Supplier Innovating Science™ Supplier No. IS8001

Includes

Materials for 15 groups: 250mL Hydrochloric Acid, 2M/2N solution UN1789 (8); 250mL Sodium Hydroxide, 2M/2N solution UN1824 (8); 500mL Ammonium Chloride, 2M solution (2); 500mL Ammonium Hydroxide, 2M solution (2); teacher's guide; and student copy masters

Witness acid-base neutralization as an exothermic process.

The heat generated by these reactions can be calculated from the mass of solution, specific heat of the solution and the change in temperature. This heat quantity can be converted to enthalpy in terms of kJ/mole by using the concentration of the reactants.

Specifications

Product Type thermochemistry Kit
Color White
Grade 7 to 14
Age 13 to 20
Includes 8 x 250ml hydrochloric acid, 2m/2n solution, 8 x 250ml sodium hydroxide, 2m/2n solution, 2 x 500ml ammonium chloride, 2m solution, 2 x 500ml ammonium hydroxide, 2m solution, teacher's guide and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 12 x 12 x 12.25 in.
Class Size 15 Lab Groups
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.