Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ ATP Muscle Set Educational Kit
GREENER_CHOICESCI_ED

Catalog No. S28492 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S28492 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S28492 Supplier Innovating Science™ Supplier No. IS3015
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

3 sets of muscles; Psoas Chemical A ( Salts); Psoas Chemical B (ATP Solution); Psoas Chemical C (Salts); 1 pack Microscope slides; 1 pack Covers slips.

Students can observe and measure how muscles contract.

  • Muscles are shipped in glycerol which extracts most of the water soluble material leaving muscle tissue that can be identified and studied using a microscope.
  • Muscle can also be stimulated to contract using the ATP chemical solution.
  • Contains enough materials for 15 groups.
  • DOT info: Nonregulated.
  • Important Notes: Chemicals must be refrigerated immediately. Muscles must also be refrigerated immediately and stored at -4°C until you are ready to use them.
  • Once refrigerated, muscle tissue has a shelf life of 4 months.

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational use
Includes 3 sets of muscles; Psoas Chemical A ( Salts); Psoas Chemical B (ATP Solution); Psoas Chemical C (Salts); Microscope slides; Cover slips.
Quantity Each
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Product Type Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.