Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Innovating Science Cellular Respiration Kit
GREENER_CHOICESCI_ED

Catalog No. s05881 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S05881 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05881 Supplier Innovating Science™ Supplier No. IS3012
Only null left
Add to Cart
Add to Cart

What Sugar Does Yeast Like Best?

  • Sugars used are glucose, sucrose, and lactose
  • Living yeast cells are exposed to sugars
  • Cells utilize sugars as food source if they are capable of metabolizing them
  • Materials for 15 groups

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Determines effectiveness of three different sugars as food source for yeast
Includes 1 x 50 mL litmus blue 0.5%, 1 x 5 g yeast instant dry, 1 x 50 g dextrose (d-glucose), 1 x 50 g sucrose, 1 x 50 g lactose monohydrate, 1 x 25 mL sodium hydroxide 0.1n
Quantity 1
Dimensions (L x W x H) 11.125 x 8.75 x 4 in.
Class Size 15 Lab Groups
Product Type Cellular Respiration Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.