Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Classification of Chemicals: Elements, Compounds and Mixtures Educational Kit
GREENER_CHOICESCI_ED

Catalog No. S28485 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S28485 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Crystal Violet and Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S28485 Supplier Innovating Science™ Supplier No. IS2513
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Crystal Violet and Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.

Three lab activities to help understand the differences between elements, compounds and mixtures. Innovating science Classification of Chemicals: Elements, Compounds and Mixtures Educational Kit, allows students to differentiate between three major classes to matter.

  • Contains enough materials for 15 groups
  • Teacher's manual and Student Study Guide copymasters are included
  • Kit Includes:
    • 0.5% Toluidine Blue (0.5 mL)
    • 0.1% Crystal Violet (0.5 mL)
    • 0.25% Safranin O (0.5 mL)
    • Chromatographic Dye Mixture (0.5 mL)
    • 95% Ethanol (400 mL)
    • 12.79g Copper Chloride EZ Prep (Makes 750 mL of 0.1M)
    • 15 Iron Metal Strips
    • 15 Chromatography Paper Sheets
    • 8 Capillary Tubes

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational use
Quantity 1 Kit
Dimensions (L x W x H) 7 x 7 x 12 in.
Class Size 15 Lab Groups
Product Type Classification of Chemicals: Elements, Compounds and Mixtures Educational Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.