Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Determination of Salinity Educational Kit
GREENER_CHOICESCI_ED

Catalog No. s28506 Shop All Aldon Products
Change view
Click to view available options
Class Size:
40 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S28506 40 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Chromium, Hexavalent Compounds, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S28506 Supplier Innovating Science™ Supplier No. IS9755
Only null left
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Chromium, Hexavalent Compounds, which are known to the State of California to cause cancer and reproductive harm.

Includes

100 mL Silver Nitrate Titrant; 15 mL 1.0M Potassium Chromate; 3 Plastic Pipettes; 3 Reaction Vessels; 3 Collection Bottles; 2 1 mL Titrators; 1 Measuring Tube

Designed to allow analyst to perform simple titration to determine the salinity of water sample. Innovating Science Determination of Salinity Educational Kit in which the titration uses an easily identifiable color transition to indicate the end of analysis.

Contains enough materials to perform 40 tests

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational use
Quantity 40 Tests
Class Size 40 Students
Product Type Determination of Salinity Educational Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.