Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Distance Learning: Gas Laws
GREENER_CHOICESCI_ED

Catalog No. S798797 Shop All Aldon Products
Change view
Click to view available options
Class Size:
2 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S798797 2 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains chemicals including phthalates, which is known to the state of CA to cause reproductive harm
Catalog No. S798797 Supplier Innovating Science™ Supplier No. IS1516
Add to Cart
Add to Cart
Contains chemicals including phthalates, which is known to the state of CA to cause reproductive harm

Students use temperature and pressure sensors to measure and plot the physical characteristics of a gas

  • Measure and plot physical characteristics of a gas (volume, pressure, temperature and number of particles)
  • Contains enough materials for two groups of students

Includes:

  • 2 gas law demonstration apparatuses
  • 2 rubber stoppers (2-hole)
  • 2 rubber stoppers (1-hole)
  • 4 styrofoam cups
  • 2 plastic test tubes
  • Instruction manual

Notes:

  • Does not include Neulog temperature and pressure sensors

Specifications

Certifications/Compliance Next gen aligned
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Chemistry
Includes 2 gas law demonstration apparatuses, 2 rubber stoppers (2-hole), 2 rubber stoppers (1-hole), 4 styrofoam cups, 2 plastic test tubes, instruction manual
Material Cardboard Box Kit
Class Size 2 Lab Groups
Product Type Characteristics of Gas Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.