Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ DNA/Chromosome Staining Kit
GREENER_CHOICESCI_ED

Catalog No. s98657 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S98657 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Methanol/ Methyl Isobutyl Ketone, which is known to the State of California to cause cancer and reproductive harm.
Catalog No. S98657 Supplier Innovating Science™ Supplier No. IS3000
Only null left
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Methanol/ Methyl Isobutyl Ketone, which is known to the State of California to cause cancer and reproductive harm.

Identify phases of plant mitosis and chromosomal development.

  • Students will be able to prepare, stain and mount slides using specially prepared onion root tips
  • Kit Includes:
    • Preserved Root Tips (1 Vial)
    • 6M Hydrochloric Acid (2 Bottles)
    • 1% Toluidine Blue Solution (2 Bottles)
    • Microscope Slides/Coverslips (1 Box)
    • 30 Aluminum Dishes
    • 30 Forceps

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Identifying the phases of plant mitosis and chromosomal development
Quantity 1 Kit
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Product Type DNA Or Chromosome Staining Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.