Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ DNA Fingerprinting Electrophoresis Lab Activity Educational Kit
GREENER_CHOICESCI_ED

Catalog No. S28493 Shop All Aldon Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S28493 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S28493 Supplier Innovating Science™ Supplier No. IS3301
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

200mL Prepared Agarose 0.8% Solution; 500mL Tris-Borate-EDTA Buffer 5X; 60mL DNA Stain; 10μg Crime Scene DNA; 10μg Victim DNA; 10μg Suspect #1 DNA; 10μg Suspect #2 DNA

Experiments to perform agarose gel electrophoresis on four DNA samples to determine if DNA from suspects matches that of DNA found at a crime scene. Innovating Science DNA Fingerprinting Electrophoresis Lab Activity Educational Kit, help students to learn about restriction enzymes and importance of these enzymes in DNA fingerprinting process.

  • Contains enough materials for 8 groups
  • Includes Teacher’s Guide and Student Study copymasters

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational use
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 8 Lab Groups
Product Type DNA Fingerprinting Electrophoresis Lab Activity Educational Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.