Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ DNA Paternity Testing Electrophoresis Lab Activity Educational Kit
GREENER_CHOICESCI_ED

Catalog No. S28494 Shop All Aldon Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S28494 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S28494 Supplier Innovating Science™ Supplier No. IS3303
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

200mL Prepared Agarose 0.8% Solution; 500mL Tris Borate EDTA Buffer 5X; 60mL DNA Stain; 10μg Mother's DNA Sample; 10μg Child's DNA Sample; 10μg Possible Father #1 DNA Sample; 10μg Possible Father #2 DNA Sample

Activity to perform electrophoresis to separate DNA samples of a child, mother, and two potential fathers to determine if either father is a possible paternal match. Innovating Science DNA Paternity Testing Electrophoresis Lab Activity Educational Kit, allows testing of child’s paternal relationship by comparing the child’s DNA profile to the profile of the mother and possible father to determine if an individual is the father or not.

  • Contains enough materials for 8 groups of students
  • Includes a Teacher’s Guide and Student Study Guide copymasters

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational use
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 8 Lab Groups
Product Type DNA Paternity Testing Electrophoresis Lab Activity Educational Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.