Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ DNA Stain (20X)
SCI_ED

Catalog No. S20654 Shop All Aldon Products
Change view
Click to view available options
Quantity:
60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S20654 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S20654 Supplier Innovating Science™ Supplier No. IS5209
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stain and Destain in 40 Minutes. This highly sensitive stain will stain a gel in about 20 minutes and destain in 20 minutes.

  • Stained gels can be stored for months without fading
  • Sufficient to stain up to 20 gels

Specifications

Color White
Concentration 1
Format Bottle
Form Liquid
Quantity 60 mL
Storage Requirements Ambient
Product Type DNA Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.