Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Exploring Meteorology
GREENER_CHOICESCI_ED

Catalog No. S40768 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S40768 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S40768 Supplier Innovating Science™ Supplier No. IS8706
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This experiment provides students with the experience of being a meteorologist.

  • Studies of pressure changes and creating a barometer
  • Studies of wind speed and creating a anemometer
  • Learn the skills to confidently read a weather map
  • Includes a demonstration of how clouds form by allowing the instructor to form a vapor cloud
  • Experiment includes material for 15 groups of 2 to perform the tasks
  • Demonstration portion has enough material to perform the demonstration 5 times
  • Aligned to the Next Generation Science Standards (NGSS)*
  • Disciplinary Core Ideas: ESS2.C; ESS2.D
  • Performance Expectations: MS- ESS2-5
  • Cross Cutting Concepts: Cause and Effect
  • Engineering Practices: Planning and Carrying Out Investigations

Specifications

Certifications/Compliance Next Gen Aligned
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Earth Science
Includes 10 mL Isopropyl Alcohol 99%; 15 Test Tubes; 15 Cork Stoppers, Small; 15 Bottles; 60 Cups, 3 oz ; 30 Cups, 5 oz ; 15 Stir Sticks; 30 Straws; 15 Balloons; 1 Dispenser; 15 Rubber Bands; 1 Glass Bottle; 15 Push Pins; 15 Practice Weather Maps
Material Cardboard Box Kit
Quantity 1
Dimensions (L x W x H) 12 x 9 x 4 in.
Class Size 15 Lab Groups
Product Type Meteorology Kit
Show More Show Less

Next Generation Science Standards is a registered trademark of Achieve. Neither Achieve nor the lead states and partners that developed the Next Generation Science Standards was involved in the production of, and does not endorse, this product.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.