Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ EZ Prep Chemicals (A-M)
SCI_ED

Catalog No. S4052A Shop All Aldon Products
Change view
Click to view available options
Product Type:
Basic Fuschin 1%
Benedicts Reagent
Bromocresol Purple 0.4%
Bromothymol Blue 0.1%
Bromothymol Blue 0.5%
Crystal Violet 1%
Eosin Y 1%
Hydrochloric Acid 1M
Malachite Green 1%
Methyl Green 1%
Methyl Orange 0.1%
Methylene Blue 0.1%
Methylene Blue 1%
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S4052A Malachite Green 1%
S96664 Benedicts Reagent
S4049A Methylene Blue 1%
S4050A Methylene Blue 0.1%
S4051A Methyl Green 1%
S4054A Eosin Y 1%
S4053A Basic Fuschin 1%
S4056A Crystal Violet 1%
S4060A Methyl Orange 0.1%
S4059A Bromothymol Blue 0.5%
S4057A Bromothymol Blue 0.1%
S4063A Bromocresol Purple 0.4%
S05890 Hydrochloric Acid 1M
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. S4052A Supplier Aldon Corporation Supplier No. IS4052
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Capsules are pre-measured to make a chemical solution.

  • Add to distilled water and stir to make the amount of solution indicated
  • There is no need to handle chemicals as the chemicals are contained within the capsule
  • Decrease preparation time since no weighing of chemicals is required
  • Decrease disposal costs because no extra chemicals remain

Specifications

Grade 7 Lab
Format Capsule
Product Type Malachite Green 1%
Quantity 1 Capsule
Color White
Languages English
For Use With (Application) Preparations of chemical solutions
Subject Chemistry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.