Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Forensic Analysis of Fabric and Fibers Educational Kit
GREENER_CHOICESCI_ED

Catalog No. S28503 Shop All Aldon Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S28503 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Ethylbenzene, which is known to the State of California to cause cancer.
Catalog No. S28503 Supplier Innovating Science™ Supplier No. IS9018
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Ethylbenzene, which is known to the State of California to cause cancer.

Includes

4 x 25mL Acetone; 4 x 25mL Sodium Hypochlorite (Bleach); 4 x 25mL Hydrochloric Acid; 4 x 25mL Sodium Hydroxide; 1 x 25mL Mounting Medium; 1 Pack pH Strips; 1 Pack Microscope Slides; 2 Packs Microscope Cover Slips; 1 Pack Lead Acetate Paper; 10 Metal Forceps; 1 Piece Cotton Fabric Sample; 1 Piece Wool Fabric Sample; 1 Piece Acetate Fabric Sample; 1 Piece Polypropylene Fabric Sample; 1 Piece Polyester Fabric Sample; 1 Piece Crime Scene Sample; 1 Piece Suspect #1 Sample; 1 Piece Suspect #2 Sample; 1 Piece Suspect #3 Sample; 1 Piece Suspect #4 Sample

Designed to introduce students to the concepts of fabric and fiber analysis. Innovating Science Forensic Analysis of Fabric and Fibers Educational Kit, cover the areas of cloth weave identification, microscopic fiber analysis, burn testing and chemical analysis.

  • Students will be given a crime scene sample and need to test materials taken from four suspects to try and identify who might be involved
  • Contains enough materials for 10 groups
  • Teachers manual and Student Study guide copymasters are included

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational use
Quantity 1
Dimensions (L x W x H) 12 x 9.25 x 4 in.
Class Size 10 Lab Groups
Product Type Forensic Analysis of Fabric and Fibers Educational Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.