Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Forensic Case Study: Drugs -Small Town, Big Problem
GREENER_CHOICESCI_ED

Catalog No. S40779 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S40779 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
WARNING: This product can expose you to chemicals including Phenolphthalein and Methanol/Methyl isobutyl ketone, which are known to the State of California to cause cancer, birth defects and reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. S40779 Supplier Innovating Science™ Supplier No. IS9024
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
WARNING: This product can expose you to chemicals including Phenolphthalein and Methanol/Methyl isobutyl ketone, which are known to the State of California to cause cancer, birth defects and reproductive harm. For more information go to www.P65Warnings.ca.gov.

In this activity students work for the Mapledale Forensic Analysis Lab and have been asked to analyze a variety of forensic evidence, including fingerprints, unknown powders, urine specimens, money, and handwriting samples, to assist the Riverside police to find a suspect.

In the small community of Riverside, police have noticed a recent increase in drug-related activity. Under orders of the mayor, the police department was tasked with solving the problem. After extensive investigation the police determined the distributor of the drugs is most likely one of six suspects. The Riverside police have asked the police department from the neighboring town of Mapledale to assist in the investigation.

  • Kit includes enough materials for 15 groups
  • Teacher's Guide and Student Analysis copymasters are included
  • Aligned to the Next Generation Science Standards (NGSS)*
  • Disciplinary Core Ideas: PS1.A; PS1.B; LS1.A
  • Performance Expectations: MS-PS1-2; MS-LS1-3
  • Cross Cutting Concepts: Patterns; Systems and System Models
  • Engineering Practices: Analyzing and Interpreting Data

Specifications

Certifications/Compliance Next Gen Aligned
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Forensics
Includes 1 x 25 g Suspect 1: White Powder Evidence; 1 x 25 g Suspect 2: White Powder Evidence; 1 x 25 g Suspect 3: White Powder Evidence; 1 x 25 g Suspect 4: White Powder Evidence; 1 x 25 g Suspect 5: White Powder Evidence; 1 x 25 g Suspect 6: White Powder Evidence; 1 x 10 mL Suspect 1: Urine Sample; 1 x 10 mL Suspect 2: Urine Sample; 1 x 10 mL Suspect 3: Urine Sample; 1 x 10 mL Suspect 4: Urine Sample; 1 x 10 mL Suspect 5: Urine Sample; 1 x 10 mL Suspect 6: Urine Sample; 3 x 10 mL Drug Detection Reagent; 3 x 10 mL Drug Extraction Reagent; 3 x 10 mL Drug Metabolite Detection Reagent; 15 strips Suspect 1: Money Evidence; 15 strips Suspect 2: Money Evidence; 15 strips Suspect 3: Money Evidence; 15 strips Suspect 4: Money Evidence; 15 strips Suspect 5: Money Evidence; 15 strips Suspect 6: Money Evidence; 15 Spot Plates
Material Cardboard Box Kit
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Product Type Forensic Kit
Show More Show Less

Next Generation Science Standards is a registered trademark of Achieve. Neither Achieve nor the lead states and partners that developed the Next Generation Science Standards was involved in the production of, and does not endorse, this product.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.