Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Forensic Case Study: Murder At Eagle Nest Harbor
GREENER_CHOICESCI_ED

Catalog No. S40780 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S40780 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S40780 Supplier Innovating Science™ Supplier No. IS9025
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A murder in a quiet house boat community sets the stage for students to test their forensic knowledge. Students will take on the role of a forensic technician as they analyze hair, fiber, fingerprint, blood, and hand writing evidence to help pin point the killer.

  • Kit provides a unique and complex set of evidence for students to analyze
  • Includes enough evidence and reagents for 15 groups to perform an analysis as well as teacher and student guide master copies
  • Aligned to the Next Generation Science Standards (NGSS)
  • Disciplinary Core Ideas: PS1.A; PS1.B
  • Performance Expectations: MS- PS1-2
  • Cross Cutting Concepts: Structure and Function
  • Engineering Practices: Developing and Using Models

Specifications

Certifications/Compliance Next Gen Aligned
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Forensics
Includes 1 x 10 mL Crime Scene: Blood Evidence; 1 x 10 mL Suspect 1: Blood Evidence; 1 x 10 mL Suspect 2: Blood Evidence; 1 x 10 mL Suspect 3: Blood Evidence; 1 x 10 mL Suspect 4: Blood Evidence; 1 x 10 mL Suspect 5: Blood Evidence; 3 x 10 mL Anti-A Antiserum; 3 x 10 mL Anti-B Antiserum; 3 x 10 mL Anti-Rh Antiserum; 15 pieces Crime Scene: Thread Evidence; 15 pieces Suspect 1: Thread Evidence; 15 pieces Suspect 2: Thread Evidence; 15 pieces Suspect 3: Thread Evidence; 15 pieces Suspect 4: Thread Evidence; 15 pieces Suspect 5: Thread Evidence; 1 bag Crime Scene: Hair Evidence; 1 bag Suspect 1: Hair Evidence; 1 bag Suspect 2: Hair Evidence; 1 bag Suspect 3: Hair Evidence; 1 bag Suspect 4: Hair Evidence; 1 bag Suspect 5: Hair Evidence; 90 Blood Typing Trays; 1 pkg Toothpicks; 1 pkg Microscope slides; 1 pkg Coverslips
Material Cardboard Box Kit
Quantity 1
Dimensions (L x W x H) 12 x 9 x 4 in.
Class Size 15 Lab Groups
Product Type Forensic Kit
Show More Show Less

Next Generation Science Standards is a registered trademark of Achieve. Neither Achieve nor the lead states and partners that developed the Next Generation Science Standards was involved in the production of, and does not endorse, this product.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.