Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Forensic Toxicology
GREENER_CHOICESCI_ED

Catalog No. S40770 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S40770 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Phenolphthalein/Methanol/Methyl isobutyl ketone, which are known to the State of California to cause cancer, birth defects and reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. S40770 Supplier Innovating Science™ Supplier No. IS9022
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Phenolphthalein/Methanol/Methyl isobutyl ketone, which are known to the State of California to cause cancer, birth defects and reproductive harm. For more information go to www.P65Warnings.ca.gov.

Students utilize different lab techniques to determine the presence of alcohol, illicit drugs and pain medications present in the simulated urine samples of suspected criminals.

  • Various tests include the Marquis Test for Morphine, Simon test for MDMA, THC Immunoassay, Cocaine Immunoassay, Marquis Test for Morphine, 6-Monoactylmorphine Test
  • Kit includes enough materials for 30 students
  • Teacher Manual and Student Study guide copy masters are included
  • Aligned to the Next Generation Science Standards (NGSS)
  • Disciplinary Core Ideas: PS1.A; PS1.B
  • Performance Expectations: MS-PS1-2; MS-PS1-3; MS-PS1-5; MS-LS1-8; HS-PS1-1; HS-PS1-2; HS-PS1-3
  • Cross Cutting Concepts: Patterns; Energy and Matter; Structure and Function
  • Engineering Practices: Obtaining, Evaluating and Communicating Information; Analyzing and Interpreting Data; Developing and Using Models; Planning and Carrying Out Investigations

Specifications

Certifications/Compliance Next Gen Aligned
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Forensics
Includes 100 mL Driver urine sample; 100 mL Passenger #1 urine sample; 100 mL Passenger #2 urine sample; 100 mL Passenger #3 urine sample; 100 mL Patient #1 urine sample; 100 mL Patient #2 urine sample; 100 mL Patient #3 urine sample; 100 mL Patient #4 urine sample; 100 mL Patient #5 urine sample; 400 mL Patient #6 urine sample; 400 mL Patient #7 urine sample; 400 mL Patient #8 urine sample; 400 mL Patient #9 urine sample; 400 mL Patient #10 urine sample; 400 mL Patient #11 urine sample; 200 mL Patient #12 urine sample; 200 mL Patient #13 urine sample; 200 mL Patient #14 urine sample; 200 mL Patient #15 urine sample; 120 mL Synthetic cocaine indicator; 30 mL Synthetic ethanol test reagent; 60 mL Synthetic marquis reagent; 30 mL Synthetic Simon’s reagent; 2 mL Ferric chloride 1.0M; 500 mL Synthetic THC indicator Solution 1; 60 mL Synthetic THC indicator Solution 2; 30 mL Synthetic 6-monoacetylmorphine; 15 Synthetic marquis reagent color charts Indicator
Material Cardboard Box Kit
Quantity 1
Dimensions (L x W x H) 10 x 10 x 10 in.
Class Size 15 Lab Groups
Product Type Toxicology Kit
Show More Show Less

Next Generation Science Standards is a registered trademark of Achieve. Neither Achieve nor the lead states and partners that developed the Next Generation Science Standards was involved in the production of, and does not endorse, this product.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.