Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Formaldehyde Solution Spill Kit Educational Kit
GREENER_CHOICESCI_ED

Catalog No. S28498 Shop All Aldon Products
Change view
Click to view available options
Class Size:
1 Class
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S28498 1 Class
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S28498 Supplier Innovating Science™ Supplier No. IS5035
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

1 Instruction Sheet/SDS; 2 Vinyl Exam Gloves; 2 9 x 16 in. Polybags; 2 Twist Ties; 2 Blank Shipping Tags; 1 Dust Pan and Brush; 1 Bag Vermiculite Absorbent; 1 kg Formaldehyde Neutralizing Agent (Sodium Bisulfite)

Contains enough material to clean up a liter of 10% formalin solution. Innovating Science Formaldehyde Solution Spill Kit Educational Kit, helps clean formaldehyde solution spills in lab during experiments.

  • Complete with instruction sheets and SDS

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational use
Includes 1 Instruction Sheet/SDS, 2 Nitrile Gloves, 2 9 x 16 in. Polybags, 2 Twist Ties, 2 Blank Shipping Tags, 1 Dust Pan and Brush, 1 Bag Vermiculite Absorbent, 1 kg Formaldehyde Neutralizing Agent (Sodium Bisulfite)
Quantity 1
Dimensions (L x W x H) 10 x 8 x 8.25 in.
Class Size 1 Class
Product Type Formaldehyde Solution Spill Kit Educational Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.