Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Instant Drosophila Medium, White
GREENER_CHOICESCI_ED

Catalog No. S28499 Shop All Aldon Products
Change view
Click to view available options
Class Size:
1 Class
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S28499 1 Class
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S28499 Supplier Innovating Science™ Supplier No. IS5036
Add to Cart
Add to Cart

To prepare instant Drosophila media just by adding water. Innovating Science Instant Drosophila Medium, White, half liter will make 30-40 standard cultures which requires no sterilization.

  • Drosophila media prepared instantly just by adding water

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational use
Includes Instant Drosophila Media, White 0.5 L
Quantity 0.5 L
Class Size 1 Class
Product Type Instant Drosophila Medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.