Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Introduction to Enzyme Catalyzed Reactions Educational Kit
GREENER_CHOICESCI_ED

Catalog No. S28496 Shop All Aldon Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S28496 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S28496 Supplier Innovating Science™ Supplier No. IS3502
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

250mL Starch Solution Quick Prep; 200mL Sucrose Solution Quick Prep; 200mL Invertase Solution Quick Prep; 150mL Amylase Solution Quick Prep; 100mL Papain Solution Quick Prep; 15mL Benedicts Qualitative Solution; 15g Gelatin Powder; 100 Graduated Plastic Pipette; 50 Student Solution Bottles with labels

Designed to introduce students to the concepts of enzymes. Innovating Science Introduction to Enzyme Catalyzed Reactions Educational Kit, helps students perform a series of four experiments involving invertase, amylase and papain, demonstrating different enzyme functions, activities and specificities.

  • Contains enough materials for 10 groups
  • Teacher’s manual and Student Study Guide copymasters are included

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational use
Quantity 1
Dimensions (L x W x H) 10 x 10 x 10 in.
Class Size 10 Lab Groups
Product Type Introduction to Enzyme Catalyzed Reactions Educational Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.