Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Kidneys and Blood Filtration
GREENER_CHOICESCI_ED

Catalog No. S98651 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S98651 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Benzidine-based dyes, which is known to the State of California to cause cancer.
Catalog No. S98651 Supplier Innovating Science™ Supplier No. IS3006
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Benzidine-based dyes, which is known to the State of California to cause cancer.

Learn the role of the kidney in blood filtration and waste removal.

  • Discover the many functional tasks performed by nephrons
  • Students will create an artificial kidney model to filter simulated blood
  • Visually determine if filtration of the simulated blood has occurred
  • Chemically test the resulting filtrate to detect any waste material removed by the kidney
  • Contains enough materials for 15 groups
  • Kit Includes:
    • Simulated unfiltered blood (100 mL)
    • Dialysis tubing (15 pieces)
    • 15 plastic cups
    • 15 pipets (1 mL)
    • 30 salt test strips
    • 30 urea test strips

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Understanding the role of the kidney in blood filtration and waste removal
Quantity 1 Kit
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Product Type Kidney and Blood Filtration Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.