Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Loading Dye (10X)
SCI_ED

Catalog No. S20653 Shop All Aldon Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S20653 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S20653 Supplier Innovating Science™ Supplier No. IS5208
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for tracking DNA during agarose electrophoresis.

Specifications

Detection Method Fluorescence
Concentration 10x
Format Bottle
Sample Type Cell Extracts, Immunoprecipitates, Purified Enzymes
For Use With (Application) Electrophoresis
For Use With (Equipment) Electrophoresis
Label or Dye Dye
Product Type Loading Dye
Purity 10X
Quantity 5 mL
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.