Learn More
Innovating Science™ Oxidation-Reduction Flag
SCI_ED
Catalog No. S96754 Shop All Aldon Products
Change view
Click to view available options
The Oxidation-Reduction Flag
Catalog No.
S96754 The Oxidation-Reduction Flag
1 options
This item is not returnable. View return policy
Catalog No. S96754 Supplier Innovating Science™ Supplier No. IS7026/DEL
This item is not returnable. View return policy
Includes
Material for five demonstrations: 25mL Iron (III) Chloride, 0.1 solution UN 2582 (3); 30mL Potassium Thiocyanate, 0.1M solution; 30mL Potassium Ferrocyanide, 0.1M solution; and instructions
Demonstrate the principles of a redox reaction.
  • Paint potassium thiocyanate and potassium ferrocyanide solutions onto filter paper
  • Once dry, spray with iron (II) chloride Watch blue and red colors appear