Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Photosynthesis: The Hill Reaction Educational Kit
GREENER_CHOICESCI_ED

Catalog No. S28495 Shop All Aldon Products
Change view
Click to view available options
Class Size:
6 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S28495 6 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S28495 Supplier Innovating Science™ Supplier No. IS3500
Add to Cart
Add to Cart

Includes

40mL 10% Propylene Glycol Solution; 500mL Plant Extract Buffer; 100mL 0.05% Dichloroindophenol EZprep; 100mL 0.5 M Sucrose EZprep; 0.25g Sodium Hydrosulfite; 2 Filters; 30 Graduated Pipettes

Activities in this kit focus on understanding the function of light cycle by running experiments based on the Hill reaction. Innovating Science Photosynthesis: The Hill Reaction Educational Kit, helps analyzing the process of converting light energy into usable chemical energy (photosynthesis).

  • Instructions are written for both visual analysis and/or UV-Vis analysis
  • Contains enough materials for 6 groups
  • Teacher’s manual and Student Study Guide copymasters are included

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational use
Quantity 1
Dimensions (L x W x H) 7 x 7 x 12 in.
Class Size 6 Lab Groups
Product Type Photosynthesis: the Hill Reaction Educational Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.