Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Principles of Stoichiometry Educational Kit
GREENER_CHOICESCI_ED

Catalog No. S28517 Shop All Aldon Products
Change view
Click to view available options
Class Size:
5 Unique Sets for 6 Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S28517 5 Unique Sets for 6 Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Lead and Lead Compounds, Chromium/ Hexavalent Compounds, soluble Nickel Compounds and Strong Inorganic Acid Mists containing Sulfuric Acid, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S28517 Supplier Innovating Science™ Supplier No. IS8060
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Lead and Lead Compounds, Chromium/ Hexavalent Compounds, soluble Nickel Compounds and Strong Inorganic Acid Mists containing Sulfuric Acid, which are known to the State of California to cause cancer and reproductive harm.

Designed to allow students to perform experiments to see the types of reactions occur when various solutions are mixed together. Innovating Science Principles of Stoichiometry Educational Kit allows students to perform four different experiments which focus on the concepts of decomposition reactions, single replacement reactions, double replacement reactions and balanced reactions.

  • Materials in this kit are packaged to minimize the need for teacher preparation providing 5 unique sets of materials for students working in groups of 6
  • Dropper topped bottles allow students to perform the array experiments without the risk for cross contamination of chemicals
  • Teacher’s Manual and Student Study Guide copymasters are included
Includes:10 x 5mL 1.0M Cupric Sulfate; 5 x 3mL 0.1M Cupric Nitrate; 5 x 3mL 0.1M Nickel Nitrate; 5 x 15mL 0.1M Lead Nitrate; 5 x 3mL 0.1M Cobalt Nitrate; 5 Bottles labeled 0.1M Nitric Acid; 5 x 3mL 0.1M Silver Nitrate; 5 x 3mL 0.1M Aluminum Nitrate; 5 x 3mL Iron (III) Nitrate; 5 x 3mL 0.1M Sodium Carbonate; 5 x 3mL 0.1M Sodium Sulfate; 5 x 3mL 0.1N Ammonium Hydroxide; 5 x 3mL 0.1M Sodium Chloride; 5 x 3mL 0.1M Ethylenediamine TetraAcetic Acid; 5 x 15mL 0.1M Sodium Iodide; 5 x 3mL 0.1M Sodium Thiocyanate; 5 x 3mL 0.1M Sodium Chromate; 5 x 3mL 0.1M Sodium Dichromate; 5 x 3mL 0.1N Sodium Hydroxide; 5 x 30mL Deionized Water; 5 x 12mL 1.0N Hydrochloric Acid; 5 Bottles labeled 1M Nitric Acid; 5 x 12mL 1.0M Sulfuric Acid; 5 x 12mL 0.1M Calcium Nitrate; 5 x 12mL 0.1M Sodium Oxalate; 65ml 1M Nitric Acid; 5pkg 9 x 0.5” Aluminum Metal Strips; 5pkg 9 x 0.5” Zinc Metal Strips; 5pkg 0.5 x 0.5” Copper Metal Strips; 5pkg 9 x 0.5” Magnesium Metal Strips; 15 Preprinted Acetate Grid Sheet; 15 Dual Well Combo Plates; 1box Wooden Dowels; 5 Forceps; 24 Carbon Electrodes; 20 Cotton String pieces

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational use
Quantity 1
Dimensions (L x W x H) 12 x 12 x 12 in.
Class Size 5 Unique Sets for 6 Groups
Product Type Principles of Stoichiometry Educational Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.