Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Properties of Shampoo
GREENER_CHOICESCI_ED

Catalog No. S96652 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S96652 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96652 Supplier Innovating Science™ Supplier No. IS9353
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tests several properties of different shampoos.

  • Learn about the role of several shampoo ingredients
  • Test several properties of different shampoos included in the kit
  • Determine the relative viscosity, pH, flash-foam formation and retention, oil emulsification and contaminant-dispersion abilities of different shampoos

Order Info

Class size: 15 groups.

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To test several properties of different shampoos
Includes 1 x 30mL Shampoo #1, 1 x 30mL shampoo #2, 1 x 30mL shampoo #3, 1 x 30mL ink, black 10%, 1 x 30mL vegetable oil, 1 universal indicator strips, 1 graduated Plastic pipettes, 1 Copper balls, disposable medicine cups 45, Polystyrene test tubes 45, Plastic cen
Quantity 1
Dimensions (L x W x H) 11.125 x 8.75 x 4 in.
Class Size 15 Lab Groups
Product Type Properties of Shampoo Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.