Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Small Group Learning: Diffusion And Osmosis
GREENER_CHOICESCI_ED

Catalog No. S40736 Shop All Aldon Products
Change view
Click to view available options
Class Size:
5 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S40736 5 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Lead and lead compounds, which are known to the State of California to cause cancer, birth defects and reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. S40736 Supplier Innovating Science™ Supplier No. IS3704SGL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Lead and lead compounds, which are known to the State of California to cause cancer, birth defects and reproductive harm. For more information go to www.P65Warnings.ca.gov.

Students study the movement of water and nutrients across a cell membrane and observe osmosis in living tissue, investigate the relationship between surface area and volume as it relates to cells and diffusion and examine the concept of molarity as related to osmotic potential and water movement.

  • Students will also be able to explain how cell size and shape affect the overall rate of nutrient intake and water elimination
  • Students will use plant tissue to determine the molarities of unknown solutions based on the direction and degree of water movement
  • Kit contains enough materials for 4 groups
  • Teacher's manual and Student Study Guide copymasters are included

Specifications

Certifications/Compliance Next Gen Aligned
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) AP Biology
Includes 25 g Agar; 8 mL Bromothymol Blue Concentrate; 25 mL 2.0 m Hydrochloric Acid; 4 Agar Block Casting Trays; 4 Plastic Cups, 5 oz. 4 Plastic Knives; 4 Plastic Stirrers; 4 Metric Rulers; 20 pc. Dialysis Tubing; 24 Plastic Cups, 7 oz.; 1 pkg Microscope Slides; 1 pkg Coverslips; 1 Cork Borer; Sucrose QuickSolution (to make 500 mL of 1.0 msolution); Sodium chloride QuickSolution (to make 500 mL of 1.0 m solution); Glucose QuickSolution (to make 500 mL of 1.0 m solution); Ovalbumin QuickSolution (to make 500 mL of 5.0% solution); Food coloring set (red, blue, yellow, green); Sucrose QuickSolution set to make:500 mL of 0.2 m Solution (Solution #3), 500 mL of 0.4 m Solution (Solution #2), 500 mL of 0.6 m Solution (Solution #5), 500 mL of 0.8 m Solution (Solution #1), 500 mL of 1.0 m Solution (Solution #4), 500 mL Distilled Water (Solution #6)
Material Cardboard Box Kit
Quantity 1
Dimensions (L x W x H) 8 x 8 x 8 in.
Class Size 5 Lab Groups
Product Type Diffusion and Osmosis Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.