Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Small Group Learning: Forensic Chemistry Of Unknown Substances
GREENER_CHOICESCI_ED

Catalog No. S40748 Shop All Aldon Products
Change view
Click to view available options
Class Size:
5 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S40748 5 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Respirable crystalline silica, which is known to the State of California to cause cancer. For more information go to www.P65Warnings.ca.gov.
Catalog No. S40748 Supplier Innovating Science™ Supplier No. IS9006SGL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Respirable crystalline silica, which is known to the State of California to cause cancer. For more information go to www.P65Warnings.ca.gov.

In this experiment, students will use observation skills, senses, and chemical tests on a series of known substances as well as two unknown substances and then attempt to identify the mystery substances based on observations and recorded data.

Often times, when collecting evidence at a crime scene, investigators may recover substances they are unable to identify in the field. Along with evidence such as fingerprints, hair, fibers, etc., there may be traces of unknown chemicals or powders left behind by the perpetrator or perpetrators. Evidence of this nature is sent to the crime lab for identification.

  • Activity includes a Teachers Manual and Student Guide and Analysis copymasters
  • Material for 5 groups
  • Aligned to the Next Generation Science Standards (NGSS)
  • Disciplinary Core Ideas: PS1.A; PS1.B
  • Performance Expectations: MS-PS1-2
  • Cross Cutting Concepts: Patterns
  • Engineering Practices: Analyzing and Interpreting Data

Specifications

Certifications/Compliance Next Gen Aligned
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Forensics
Includes 10 Reaction Plates; 10 g Baking Powder; 10 g Baking Soda; 10 g Corn Starch; 10 g Talcum Powder; 10 g Salt; 10 g Gelatin; 10 g Mystery Substance #1; 10 g Mystery Substance #2; 10 mL Biuret Reagent; 10 mL Dilute Lugol’s Iodine; 10 mL Acetic Acid
Material Cardboard Box Kit
Quantity 1
Dimensions (L x W x H) 9 x 6 x 4 in.
Class Size 5 Lab Groups
Product Type Forensic Kit
Show More Show Less

WARNING: This product can expose you to chemicals including Respirable Crystalline Silica, which is known to the State of California to cause cancer.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.