Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Small Group Learning: Forensics Of Hair Analysis
GREENER_CHOICESCI_ED

Catalog No. S40749 Shop All Aldon Products
Change view
Click to view available options
Class Size:
5 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S40749 5 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S40749 Supplier Innovating Science™ Supplier No. IS9007SGL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Discover the differences between human and animal hair, differences among different types of human hair, and the ways forensic scientists use hair to assist in solving crimes.

  • In the second part of the activity, students will try to determine the origin of a hair sample from a crime scene in relation to hair samples from four known suspects
  • Activity includes a Teachers Manual and Student Guide and Analysis copy-masters
  • Material for 5 groups
  • Aligned to the Next Generation Science Standards (NGSS)
  • Disciplinary Core Ideas: LS1.A; LS1.C
  • Performance Expectations: HS-LS1-1; HS-LS1-6
  • Cross Cutting Concepts: Energy and Matter; Structure and Function
  • Engineering Practices: Constructing Explanations and Designing Solutions

Specifications

Certifications/Compliance Next Gen Aligned
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Forensics
Includes 1 box Microscope Slides; 1 pkg Coverslips; 1 Deer Hair Sample; 1 Cat Hair Sample; 4 Human Hair Samples; 5 Forceps
Material Cardboard Box Kit
Quantity 1
Dimensions (L x W x H) 9 x 6 x 4 in.
Class Size 5 Lab Groups
Product Type Forensic Kit
Show More Show Less

Next Generation Science Standards is a registered trademark of Achieve. Neither Achieve nor the lead states and partners that developed the Next Generation Science Standards was involved in the production of, and does not endorse, this product.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.