Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Small Group Learning: Properties Of Aspirin
GREENER_CHOICESCI_ED

Catalog No. S40754 Shop All Aldon Products
Change view
Click to view available options
Class Size:
5 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S40754 5 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Acetyl Salicylic acid/Aspirin and Phenolphthalein/Methanol/Methyl isobutyl ketone, which are known to the State of California to cause cancer, birth defects and reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. S40754 Supplier Innovating Science™ Supplier No. IS9354SGL
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Acetyl Salicylic acid/Aspirin and Phenolphthalein/Methanol/Methyl isobutyl ketone, which are known to the State of California to cause cancer, birth defects and reproductive harm. For more information go to www.P65Warnings.ca.gov.

Students will examine the solubility of aspirin in varying gastrointestinal environments, a comparison of active ingredients in regular and extra strength aspirin, and the differences between regular and buffered aspirin.

Aspirin is to this day the most widely used painkiller and largest selling non-prescription medicine in the world. Learn about the history of the development acetylsalicylic acid (aspirin) and test the performance of several different types of aspirin.

  • All aspirin samples are included
  • Kit contains enough materials for 5 groups
  • Teacher's Manual and Student Study Guide copymasters are included
  • Aligned to the Next Generation Science Standards (NGSS)
  • Disciplinary Core Ideas: PS1.A; PS1.B
  • Performance Expectations: MS-PS1-2; MS-PS1-3
  • Cross Cutting Concepts: Patterns; Structure and Function
  • Engineering Practices: Analyzing and Interpreting Data; Obtaining, Evaluating and Communicating Information

Specifications

Certifications/Compliance Next Gen Aligned
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Consumer Chemistry
Includes 1 x 50 mL Hydrochloric Acid, 1M Solution; 3 x 30 mL Sodium Hydroxide, 1M Solution; 1 x 10 mL Phenolphthalein, 1% in Ethanol; 1 Pkg/18 Universal Indicator Strips; 1 Pkg/5 Graduated Plastic Pipettes; 15 Disposable Medicine Cups; 1 Pkg/25 Aspirin, Regular; 1 Pkg/20 Aspirin, Buffered; 1 Pkg/15 Aspirin, Enteric-coated; 1 Pkg/5 Aspirin, Extra Strength
Material Cardboard Box Kit
Quantity 1
Dimensions (L x W x H) 9 x 6 x 4 in.
Class Size 5 Lab Groups
Product Type Chemistry Kit
Show More Show Less

WARNING: This product can expose you to chemicals including Acetyl Salicylic Acid/ Aspirin, and Phenolphthalein/ Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.