Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Small Group Learning: Separation Of Mixture Of Solid
GREENER_CHOICESCI_ED

Catalog No. S40731 Shop All Aldon Products
Change view
Click to view available options
Class Size:
5 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S40731 5 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S40731 Supplier Innovating Science™ Supplier No. IS2523SGL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Performing liquid-liquid extractions allows students to apply their knowledge of the properties of molecules. Students will learn how the properties of solubility can be utilized to separate two solids in a mixture.

  • Kit provides for a qualitative test that will help students visualize that two different solids were collected, despite their similar appearance.
  • Students will also explore the concept of the law of conservation of mass as well as how acid-base reactions can be utilized to manipulate molecules.
  • Kit contains enough material for 5 groups
  • Teacher's Manual and Student Study Guide copymasters are included
  • Aligned to the Next Generation Science Standards (NGSS)
  • Disciplinary Core Ideas: PS1A; PS1B
  • Performance Expectations: MS-PS1-1; MS-PS1-2; MS-PS1-5; HS-PS1-2; HS-PS1-3; HS-PS1-7
  • Cross Cutting Concepts: Scale, Proportion and Quantity; Patterns; Energy and Matter; Scientific Knowledge Assumes an Order and Consistency in Natural Systems
  • Engineering Practices: Developing and Using Models; Analyzing and Interpreting Data; Planning and Carrying Out Investigations; Scientific Knowledge is Based on
  • Empirical Evidence; Science Models, Laws, Mechanisms, and Theories Explain Natural Phenomena; Constructing Explanations and Designing Solutions; Using Mathematics and Computational Thinking

Specifications

Certifications/Compliance Next Gen Aligned
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Chemistry
Includes 15 g Benzoic Acid/Acetanilide 1:1 Mixture; 15 g Benzoic Acid/Acetanilide 2:1 Mixture; 15 g Benzoic Acid/Acetanilide 3:1 Mixture; 1 g Methyl Violet; 135 mL Ethyl Acetate; 75 mL Sodium Bicarbonate, 1.0M; 100 mL Hydrochloric Acid, 1.0N; 12 Pipettes
Material Cardboard Box Kit
Quantity 1
Dimensions (L x W x H) 8 x 8 x 8 in.
Class Size 5 Lab Groups
Product Type Chemistry Education Kit
Show More Show Less

Next Generation Science Standards is a registered trademark of Achieve. Neither Achieve nor the lead states and partners that developed the Next Generation Science Standards was involved in the production of, and does not endorse, this product.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.