Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Tris-EDTA (TE) Solution (10X)
SCI_ED

Catalog No. s20650 Shop All Aldon Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S20650 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S20650 Supplier Innovating Science™ Supplier No. IS5207
Only null left
Add to Cart
Add to Cart

Use for diluting DNA samples

TE buffer is a buffer solution commonly used in molecular biology, especially in procedures involving DNA, cDNA or RNA. The purpose of this TE buffer is for diluting DNA samples, while protecting it from degradation.

Specifications

Buffer TBE
Content And Storage Ambient
Concentration 1
Format Bottle
Sample Type Cell Extracts, Immunoprecipitates, Purified Enzymes
For Use With (Application) Microbiology
Includes Tris-EDTA (TE) Solution 10X concentrate, 25 mL
Label or Dye No
Product Type Biotechnology Reagent
Purity 10x
Quantity 25 mL
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.