Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Urinalysis Using Simulated Urine
GREENER_CHOICESCI_ED

Catalog No. S98668A Shop All Aldon Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S98668A 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S98668A Supplier Innovating Science™ Supplier No. IS3008
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Teacher's guide, student study guide copymasters, kit with simulated urine samples (250mL each of patient X, patient Y, patient Z, control), Benedict's reagent (25mL), biuret solution (25mL), Sulkowitch's reagent (25mL), pH test strips, microscope slides, coverslips, graduated plastic cups (30mL), graduated pipets (1mL)

Requires

Hot water bath, glass test tubes and compound microscopes (4X/10X/40X)

One of the oldest medical diagnostic tests performed.

Students will:

  • Use simulated urine to avoid the unpleasantness of using the real thing
  • Examine simulated urine factors such as pH, color and clarity, as well as test for the presence or absence of proteins, glucose and calcium
  • Examine samples microscopically to determine if any crystals are present in the sample

Order Info

Materials for 15 groups.

Compliance

This package conforms to 49CFR 173.4

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Use simulated urine to avoid the unpleasantness of using the real thing, examine simulated urine factors such as ph, color and clarity, as well as test for the presence or absence of proteins, glucose and calcium
Includes Teacher's guide, student study guide copymasters, kit with simulated urine samples (250mL each of patient X, patient Y, patient Z, control), 25mL Benedict's reagent, 25mL biuret solution, 25mL Sulkowitch's reagent, pH test strips, microscope slides, coverslips, 30mL graduated Plastic cups, 1mL graduated pipets
Quantity 1 Ea.
Dimensions (L x W x H) 12.125 x 9.25 x 6.5 in.
Class Size 15 Lab Groups
Product Type Urinalysis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.