Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Water Testing Kits
GREENER_CHOICESCI_ED

Catalog No. S12837 Shop All Aldon Products
Change view
Click to view available options
For Use With (Application):
Determination of Nitrate and Nitrite
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S12837 Determination of Nitrate and Nitrite
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals known to the State of California to cause and reproductive harm.
Catalog No. S12837 Supplier Innovating Science™ Supplier No. IS9753
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals known to the State of California to cause and reproductive harm.

Includes

  • Complete with instructions
  • For 40 tests

Contains everything an analyst would need to perform a water analysis either in the laboratory setting or out in the field.

  • Parameters tested:
  • Dissolved Oxygen
  • Water Hardness
  • Water's Alkalinity
  • Nitrate and Nitrite
  • Ammonia

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Determination of Nitrate and Nitrite
Includes Instructions
Quantity 40 Tests
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 40 Students
Product Type Water Testing Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.