Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Who Stole the Gold? A Comprehensive Forensic Analysis Educational Kit
GREENER_CHOICESCI_ED

Catalog No. S28504 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S28504 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Respirable Crystalline Silica and Phenolphthalein/ Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cancer and reproductive harm.
Catalog No. S28504 Supplier Innovating Science™ Supplier No. IS9019
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Respirable Crystalline Silica and Phenolphthalein/ Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cancer and reproductive harm.

To help figure out which of the four suspects is responsible for stealing the gold. Innovating Science Who Stole the Gold? A Comprehensive Forensic Analysis Educational Kit in which the experiments are carried out using techniques from soil and mineral analysis, document analysis, fabric and fiber analysis, blood analysis, fingerprint analysis and hair analysis.

  • Contains enough materials for 15 groups
  • Teacher’s Gudie and Student Study Guide copymasters included
Includes: 3 x 3mL Crime Scene Blood Sample; 3 x 3mL Suspect #1 Blood Sample; 3 x 3mL Suspect #2; Blood Sample; 3 x 3mL Suspect #3 Blood Sample; 3 x 3mL Suspect #4 Blood Sample; 3 x 5mL Simulated Anti-B Serum; 3 x 5mL Simulated Anti-A Serum; 3 x 3mL 0.5M Hydrochloric Acid; 3 x 2mL Universal Indicator; 24g Crime Scene Soil Sample; 24g Suspect #1 Soil Sample; 24g Suspect #2 Soil Sample; 24g Suspect #3 Soil Sample; 24g Suspect #4 Soil Sample; 3 Universal Indicator Color Charts; 5 Plastic Scoops; 3 Small Mesh Screens; 3 Large Mesh Screens; 15 Blood Typing Trays; 2 Boxes Microscope Slides; 1 Pack Microscope Slide Cover Slips; 3 Hand Magnifiers; 1 Pack pH Strips; 1 Pack Lead Acetate Paper; 3 Metal Forceps; 1 Crime Scene Fabric Sample; 1 Suspect #1 Fabric Sample; 1 Suspect #2 Fabric Sample; 1 Suspect #3 Fabric Sample; 1 Suspect #4 Fabric Sample; 3 Crime Scene Note; 3 Suspect #1 Writing Sample; 3 Suspect #2 Writing Sample; 3 Suspect #3 Writing Sample; 3 Suspect #4 Writing Sample; 1 Crime Scene Hair Sample; 1 Suspect #1 Hair Sample; 1 Suspect #2 Hair Sample; 1 Suspect #3 Hair Sample; 1 Suspect #4 Hair Sample

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational use
Quantity 1
Dimensions (L x W x H) 13 x 10 x 7 in.
Class Size 15 Lab Groups
Product Type Who Stole the Gold? A Comprehensive Forensic Analysis Educational Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.