Learn More
Med Vet International Innovation Microscope, Trinoc 4 Objective Scope w/ BioView Camera, Screen & 100x DRY Updgrade, J0334IT4XD, Each
Catalog No. 5000585478
Encompass_Preferred
The exceptional Innovation microscope is the top choice for discerning doctors, cytologists, and researchers, equalling the features and quality of popular Nikon and Olympus models. The infinity PLAN flat-field objectives paired with the 10x/22 super-wide high-point eyepieces produce the highest resolution over a broad field of view. It's ergonomic design and smooth rackless stage provides hours of comfort and ease of use. Powerful performance, crisp clarity, and rugged construction, backed by a lifetime warranty on optical and mechanical components make the Innovation a valued addition to any veterinary laboratory. Includes the BioVIEW HD Camera and Monitor to the Innovation microscope for live presentations and client education...it's all included in one part number! The HD camera plus 1080p 11.6" monitor provides a bright and beautiful image, with high sensitivity and ultra-fast motion.
Catalog No. 50-005-85478 Supplier Med Vet International Supplier No. JOR334IT4XD
The exceptional Innovation microscope is the top choice for discerning doctors, cytologists, and researchers, equalling the features and quality of popular Nikon and Olympus models. The infinity PLAN flat-field objectives paired with the 10x/22 super-wide high-point eyepieces produce the highest resolution over a broad field of view. It's ergonomic design and smooth rackless stage provides hours of comfort and ease of use. Powerful performance, crisp clarity, and rugged construction, backed by a lifetime warranty on optical and mechanical components make the Innovation a valued addition to any veterinary laboratory. Includes the BioVIEW HD Camera and Monitor to the Innovation microscope for live presentations and client education...it's all included in one part number! The HD camera plus 1080p 11.6" monitor provides a bright and beautiful image, with high sensitivity and ultra-fast motion.