Learn More
Chrom Matrix Inc INNOVATION STBL AMIDE 3UM CLMN
Catalog No. NC9896578
Encompass
Rugged, long-lived amide HPLC column. High resolution, fast analysis and no column carryover. Excellent peak shapes for difficult basic compounds.
Catalog No. NC9896578 Supplier Chrom Matrix Inc Supplier No. 5732110U
Rugged, long-lived amide HPLC column. High resolution, fast analysis and no column carryover. Excellent peak shapes for difficult basic compounds.