Learn More
Innovative Research Inc Innovative Grade US Origin Mouse Balb C Serum, 10 ml, Serum, Whole blood collection, Balb C mouse
Catalog No. NC2740297
Encompass_Preferred
Innovative Research Inc.'s Innovative Grade US Origin Mouse Balb C Serum is a premium biological reagent designed to provide researchers with a highly consistent and reliable material crucial for various scientific applications. This serum is sourced from whole blood collected from Balb C mice of US origin and ensures high purity while delivering consistent performance for sensitive experiments.
  • High purity and quality sourced from US Origin Balb C mice.
  • Consistent performance processed for dependable and reproducible results.
  • Specific strain sourcing ideal for studies in immunology and cancer research.
  • Volume of 10 ml.
  • Whole blood collection method.
  • Storage recommendation: Use immediately or store at 4 °C for up to 24 hours after thawing.
  • Freezing warning: Do not keep on ice or store below 0 °C to prevent clotting.
Catalog No. NC2740297 Supplier Innovative Research Inc Supplier No. IGMSBCSER10ML
Innovative Research Inc.'s Innovative Grade US Origin Mouse Balb C Serum is a premium biological reagent designed to provide researchers with a highly consistent and reliable material crucial for various scientific applications. This serum is sourced from whole blood collected from Balb C mice of US origin and ensures high purity while delivering consistent performance for sensitive experiments.
  • High purity and quality sourced from US Origin Balb C mice.
  • Consistent performance processed for dependable and reproducible results.
  • Specific strain sourcing ideal for studies in immunology and cancer research.
  • Volume of 10 ml.
  • Whole blood collection method.
  • Storage recommendation: Use immediately or store at 4 °C for up to 24 hours after thawing.
  • Freezing warning: Do not keep on ice or store below 0 °C to prevent clotting.