Learn More
Innovative Reseach Of America INNOVATIVE RESEARCH OF AMERICA 17β-Estradiol Pellet, 0.18 mg, 60-Day Release
SDP
Catalog No. NC3984771
Encompass
This product is a 17β-Estradiol pellet designed for slow, sustained release. It contains 0.18 mg of 17β-Estradiol and provides a 60-day release profile, making it suitable for studies requiring consistent hormone delivery, such as estrogen replacement or tumor growth induction.
  • Contains 0.18 mg of 17β-Estradiol.
  • Provides a 60-day sustained release.
  • Designed for subcutaneous implantation.
  • Supports research in estrogen replacement.
  • Useful for studies involving tumor growth induction in models like MCF-7 cell line implanted SCID-SHO mice.
Catalog No. NC3984771 Supplier Innovative Reseach Of America Supplier No. NE12190D0.18MG50
This product is a 17β-Estradiol pellet designed for slow, sustained release. It contains 0.18 mg of 17β-Estradiol and provides a 60-day release profile, making it suitable for studies requiring consistent hormone delivery, such as estrogen replacement or tumor growth induction.
  • Contains 0.18 mg of 17β-Estradiol.
  • Provides a 60-day sustained release.
  • Designed for subcutaneous implantation.
  • Supports research in estrogen replacement.
  • Useful for studies involving tumor growth induction in models like MCF-7 cell line implanted SCID-SHO mice.