Learn More
MilliporeSigma™ InnoZyme™ Gelatinase (MMP-2/MMP-9) Activity Assay kit, Fluorogenic
Catalog No. CBA0031KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1kit
Catalog No. Quantity
CBA0031KIT 1kit
1 options
Catalog No. CBA0031KIT Supplier MilliporeSigma™ Supplier No. CBA0031KIT
Add to Cart
Add to Cart
Useful for screening of gelatinase inhibitors
  • Sensitive fluorogenic assay (Exc. Max: ∽325 nm; Emi. Max.: ∽393 nm) for measurement of gelatinases (MMP-2 and MMP-9)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Gelatinase activity assay kit
Kit Contents Gelatinase Substrate, pro-MMP-2 Positive Control, Inhibitor, APMA, Assay Buffer, 96-Well Plate, Plate Sealer, and a user protocol
Quantity 1kit
Storage Requirements -20°C