Learn More
Medchemexpress LLC INO-1001 | 501364-82-5 | C23H25N3O4S | 100 MG
SDP
Catalog No. 5000374776
Encompass_Preferred
INO-1001 is a potent and selective poly (ADP-ribose) polymerase (PARP) inhibitor. It enhances radiation sensitivity and radiation-induced cell killing by interfering with DNA repair mechanisms, leading to necrotic cell death. This compound also exhibits anti-tumor effects and has a purity of 98.86%.
  • Potent and selective PARP inhibitor
  • Enhances radiation sensitivity
  • Interferes with DNA repair mechanisms
  • Leads to necrotic cell death
  • Exhibits anti-tumor effects
  • Purity of 98.86%
  • IC50 of 0.05 µM in Chinese hamster ovary AA8 (CHO) cells
Catalog No. 50-003-74776 Supplier Medchemexpress LLC Supplier No. HY15045100MG
INO-1001 is a potent and selective poly (ADP-ribose) polymerase (PARP) inhibitor. It enhances radiation sensitivity and radiation-induced cell killing by interfering with DNA repair mechanisms, leading to necrotic cell death. This compound also exhibits anti-tumor effects and has a purity of 98.86%.
  • Potent and selective PARP inhibitor
  • Enhances radiation sensitivity
  • Interferes with DNA repair mechanisms
  • Leads to necrotic cell death
  • Exhibits anti-tumor effects
  • Purity of 98.86%
  • IC50 of 0.05 µM in Chinese hamster ovary AA8 (CHO) cells