Learn More
Medchemexpress LLC Ino-1001 | 501364-82-5 | C23H25N3O4S | 50 MG
SDP
Catalog No. 5000374836
Encompass_Preferred
INO-1001 is a potent and selective poly (ADP-ribose) polymerase (PARP) inhibitor. It acts as a potent enhancer of radiation sensitivity, augmenting radiation-induced cell killing by interfering with DNA repair mechanisms, which leads to necrotic cell death. INO-1001 has demonstrated anti-tumor effects.
  • Potent and selective poly (ADP-ribose) polymerase (PARP) inhibitor.
  • Enhances radiation sensitivity.
  • Augments radiation-induced cell killing.
  • Interferes with DNA repair mechanisms.
  • Leads to necrotic cell death.
  • Exhibits anti-tumor effects.
  • Inhibits PARP at 0.05 μM in Chinese hamster ovary AA8 (CHO) cells.
  • ED50 of 1 μM for inhibition of PARP1 in CHO-K1 cells.
Catalog No. 50-003-74836 Supplier Medchemexpress LLC Supplier No. HY1504550MG
INO-1001 is a potent and selective poly (ADP-ribose) polymerase (PARP) inhibitor. It acts as a potent enhancer of radiation sensitivity, augmenting radiation-induced cell killing by interfering with DNA repair mechanisms, which leads to necrotic cell death. INO-1001 has demonstrated anti-tumor effects.
  • Potent and selective poly (ADP-ribose) polymerase (PARP) inhibitor.
  • Enhances radiation sensitivity.
  • Augments radiation-induced cell killing.
  • Interferes with DNA repair mechanisms.
  • Leads to necrotic cell death.
  • Exhibits anti-tumor effects.
  • Inhibits PARP at 0.05 μM in Chinese hamster ovary AA8 (CHO) cells.
  • ED50 of 1 μM for inhibition of PARP1 in CHO-K1 cells.