Learn More
Medchemexpress LLC INO-1001 | 501364-82-5 | MFCD19443171 | 98.9% | 439.53 | C23H25N3O4S | 10 MG
SDP
Catalog No. 5000286002
Encompass_Preferred
INO-1001 is a small-molecule poly(ADP-ribose) polymerase (PARP) inhibitor used in biochemical and cellular research to study DNA repair pathways and to sensitize cells to ionizing radiation. Supplied as a powder for research applications, it is characterized by CAS 501364-82-5 and has the formula C23H25N3O4S (MW 439.53).
  • Selective PARP inhibition for DNA repair studies.
  • Enhances radiosensitivity in cellular assays.
  • Suitable for biochemical and cell-based assays.
  • Provided as a powder for flexible formulation.
  • Stable when stored under recommended conditions.
  • High purity by RP-HPLC (98.86%).
Catalog No. 50-002-86002 Supplier Medchemexpress LLC Supplier No. HY1504510MG
INO-1001 is a small-molecule poly(ADP-ribose) polymerase (PARP) inhibitor used in biochemical and cellular research to study DNA repair pathways and to sensitize cells to ionizing radiation. Supplied as a powder for research applications, it is characterized by CAS 501364-82-5 and has the formula C23H25N3O4S (MW 439.53).
  • Selective PARP inhibition for DNA repair studies.
  • Enhances radiosensitivity in cellular assays.
  • Suitable for biochemical and cell-based assays.
  • Provided as a powder for flexible formulation.
  • Stable when stored under recommended conditions.
  • High purity by RP-HPLC (98.86%).