Learn More
Medchemexpress LLC HY-15045 5mg Medchemexpress, INO-1001 CAS:501364-82-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY150455MG
Encompass_Preferred
Medchemexpress, HY-15045 5mg INO-1001 CAS:501364-82-5 INO-1001 is a potent and selective Poly (ADP-ribose) polymerase (PARP) inhibitor with an IC 50 of 0.05-1 μM. INO-1001 is a potent enhancer of radiation sensitivity and enhances radiation-induced cell killing by interfering with DNA repair mechanisms, resulting in necrotic cell death [1] . INO-1001 has anti-tumor effects [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2721