Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ INO80 Recombinant Enzymes
SDP

Catalog No. NBP348874
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP348874 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP348874 Supplier Novus Biologicals™ Supplier No. NBP348874

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Recombinant Enzyme

Specifications

Gene ID (Entrez) 54617
Species Human
Purity Immunogen affinity purified
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias EC 3.6.1, EC 3.6.1.23, EC 3.6.4.12, hINO80INO80 complex subunit A, homolog of yeast INO80, Ino80, INO80 complex homolog 1 (S. cerevisiae), INO80 homolog (S. cerevisiae), INO80A, INOC1putative DNA helicase INO80 complex homolog 1, KIAA1259
Gene Symbol INO80
Quantity 50 μL
Storage Buffer 20 mM Tris-HCl, pH 7.9, 150 mM NaCl, 15% glycerol, 2 mM MgCl2, 0.2 mM EDTA, 1 mM DTT, 10 mM beta-glycerophospate, 0.4 mM PMSF, 1 mM benzamidine and 0.4 mg/mL recombinant human insulin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.