Learn More
Aviva Systems Biology Corp Ino80b Antibody - C-terminal region (ARP60851_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60851_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ino80b. It was validated on Western Blot. . Alternate names: 2510009I23Rik, HMG1YL4, Hmga1l4, Papa1, Znhit4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-188