Learn More
TARGETMOL CHEMICALS INC INOBRODIB 5MG
SDP
Catalog No. 502581685
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Inobrodib (CBP-IN-1) is a potent inhibitor of p300/CBP bromodomain. purity: 97%
Catalog No. 50-258-1685 Supplier TARGETMOL CHEMICALS INC Supplier No. T107175MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Inobrodib (CBP-IN-1) is a potent inhibitor of p300/CBP bromodomain. purity: 97%