Learn More
Cole-Parmer Inoculating loop, platinum, 2 mm ID, 24 gauge, 45 mm length
Catalog No. NC0429993 Shop All Cole-Parmer Products
Encompass_Preferred
Cole-Parmer Inoculating loop, platinum, 2 mm ID, 24 gauge, 45 mm lengthCalibrated loops 01850-00 and 01850-02 are ideal for milk samples. The 1-microliter loops are made of 26-ga B&S platinum wire with 15% iridium. The 10-microliter loops are calibrated and made of 19-ga B&S platinum wire with 5% rhodium. Order insulated loop and needle holder separately.Platinum loops and needles are used for transferring bacterial cultlres. These loops and needles are made of B&S platinum wire with 15% iridium added for rigidity, permitting a thinner wire for delicate work. Loops are fused to form a "perfect circle" with an inside tolerance of +/-0.02 mm to ensure accurate delivery. Order insulated loop and needle holder separetly.Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Catalog No. NC0429993 Supplier Cole-Parmer Supplier No. 0185004
Cole-Parmer Inoculating loop, platinum, 2 mm ID, 24 gauge, 45 mm lengthCalibrated loops 01850-00 and 01850-02 are ideal for milk samples. The 1-microliter loops are made of 26-ga B&S platinum wire with 15% iridium. The 10-microliter loops are calibrated and made of 19-ga B&S platinum wire with 5% rhodium. Order insulated loop and needle holder separately.Platinum loops and needles are used for transferring bacterial cultlres. These loops and needles are made of B&S platinum wire with 15% iridium added for rigidity, permitting a thinner wire for delicate work. Loops are fused to form a "perfect circle" with an inside tolerance of +/-0.02 mm to ensure accurate delivery. Order insulated loop and needle holder separetly.Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.